Assay Details
Gene Symbol: LOC711104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108444.4 | XP_001108444.1 | 740 | 120 | ||
XM_015135024.2 | XP_014990510.1 | 413 | 120 | |||
XM_015135025.2 | XP_014990511.1 | 745 | 120 | |||
XM_015135026.2 | XP_014990512.1 | 3095 | 120 | |||
XM_015135028.2 | XP_014990514.1 | 2615 | 120 | |||
XM_015135034.2 | XP_014990520.1 | 674 | 120 | |||
XM_015135036.2 | XP_014990522.1 | 4199 | 120 | |||
XM_015135040.2 | XP_014990526.1 | 810 | 120 | |||
XM_028846365.1 | XP_028702198.1 | 1429 | 120 | |||
GenBank mRNA | JU319762.1 | - | 627 | 120 | ||
JU471676.1 | - | 388 | 120 | |||
JV044892.1 | - | 546 | 120 |

More Information
Set Membership: |
![]() ![]() |
