Assay Details
| Gene Symbol: LSM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122421.2 | XP_014977907.1 | 158 | 103 | ||
| GenBank mRNA | JU475969.1 | - | 125 | 103 | ||
| JV043572.1 | - | 141 | 103 | |||
| JV601851.1 | - | 106 | 103 | |||
More Information
Set Membership: |
|

LSM7
Genomic Map