Assay Details
| Gene Symbol: EGFL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194009.2 | NP_001180938.2 | 309 | 101 | ||
| XM_015116317.2 | XP_014971803.2 | 640 | 101 | |||
| XM_028834341.1 | XP_028690174.1 | 691 | 101 | |||
| XM_028834342.1 | XP_028690175.1 | 368 | 101 | |||
| GenBank mRNA | JU334815.1 | - | 455 | 101 | ||
| JV668620.1 | - | 627 | 101 | |||
More Information
Set Membership: |
|

EGFL7
Genomic Map