Assay Details
| Gene Symbol: RAB23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261741.1 | NP_001248670.1 | 333 | 136 | ||
| XM_015136548.2 | XP_014992034.1 | 594 | 136 | |||
| XM_015136549.2 | XP_014992035.1 | 422 | 136 | |||
| XM_015136550.2 | XP_014992036.1 | 464 | 136 | |||
| GenBank mRNA | JU321798.1 | - | 312 | 136 | ||
| JU475972.1 | - | 333 | 136 | |||
More Information
Set Membership: |
|

RAB23
Genomic Map