Assay Details
| Gene Symbol: MSRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346686.2 | NP_001333615.2 | 372 | 76 | ||
| GenBank mRNA | JU332293.1 | - | 312 | 76 | ||
| JU473184.1 | - | 344 | 76 | |||
More Information
Set Membership: |
|

MSRB1
Genomic Map