Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194620.2 | NP_001181549.2 | 149 | 136 | ||
| XM_015117187.2 | XP_014972673.2 | 158 | 136 | |||
| GenBank mRNA | JU475924.1 | - | 139 | 136 | ||
More Information
Set Membership: |
|

TRMO
Genomic Map