Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194620.2 | NP_001181549.2 | 323 | 145 | ||
| XM_015117187.2 | XP_014972673.2 | 332 | 145 | |||
| XM_015117189.2 | XP_014972675.2 | 408 | 145 | |||
| GenBank mRNA | JU334916.1 | - | 299 | 145 | ||
| JU475924.1 | - | 313 | 145 | |||
| JV047320.1 | - | 273 | 145 | |||
More Information
Set Membership: |
|

TRMO
Genomic Map