Assay Details
| Gene Symbol: KLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261280.1 | NP_001248209.1 | 872 | 78 | ||
| XM_015138149.2 | XP_014993635.1 | 5006 | 78 | |||
| XM_015138150.2 | XP_014993636.1 | 757 | 78 | |||
| XM_015138151.2 | XP_014993637.1 | 795 | 78 | |||
| GenBank mRNA | JU334842.1 | - | 785 | 78 | ||
| JU472395.1 | - | 885 | 78 | |||
| JV047272.1 | - | 872 | 78 | |||
| JV686101.1 | - | 1032 | 78 | |||
More Information
Set Membership: |
|

KLF3
Genomic Map