Assay Details
| Gene Symbol: GPR83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265726.1 | NP_001252655.1 | 825 | 91 | ||
| XM_015115446.2 | XP_014970932.1 | 766 | 91 | |||
| GenBank mRNA | JU326335.1 | - | 643 | 91 | ||
| JV565012.1 | - | 825 | 91 | |||
| JV705634.1 | - | 494 | 91 | |||
More Information
Set Membership: |
|

GPR83
Genomic Map