Assay Details
| Gene Symbol: PLAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258163.1 | NP_001245092.1 | 315 | 133 | ||
| XM_015138310.2 | XP_014993796.1 | 386 | 133 | |||
| GenBank mRNA | JU334857.1 | - | 298 | 133 | ||
| JU475880.1 | - | 315 | 133 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLAC8
Genomic Map