Assay Details
| Gene Symbol: CKLF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265912.1 | NP_001252841.1 | 313 | 150 | ||
| XM_015126320.2 | XP_014981806.1 | 512 | 150 | |||
| GenBank mRNA | JU471340.1 | - | 313 | 150 | ||
| JU471342.1 | - | 472 | 150 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CKLF
Genomic Map