Assay Details
| Gene Symbol: POLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257857.1 | NP_001244786.1 | 189 | 112 | ||
| GenBank mRNA | JU320645.1 | - | 189 | 112 | ||
| JU320646.1 | - | 189 | 112 | |||
| JU470579.1 | - | 187 | 112 | |||
| JU470580.1 | - | 187 | 112 | |||
| JV043795.1 | - | 195 | 112 | |||
| JV043796.1 | - | 195 | 112 | |||
| JV638584.1 | - | 195 | 112 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POLE3
Genomic Map