Assay Details
| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028843921.1 | XP_028699754.1 | 1376 | 152 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028843921.1 transcripts.
Set Membership: |
|

CRYGS
Genomic Map