Assay Details
| Gene Symbol: TRIM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260523.1 | NP_001247452.1 | 1055 | 127 | ||
| XM_028832806.1 | XP_028688639.1 | 1162 | 127 | |||
| XM_028832807.1 | XP_028688640.1 | 1162 | 127 | |||
| GenBank mRNA | JU476111.1 | - | 1049 | 127 | ||
| JV044348.1 | - | 1055 | 127 | |||
More Information
Set Membership: |
|

TRIM44
Genomic Map