Assay Details
| Gene Symbol: ITLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801868.3 | XP_002801914.2 | 432 | 153 | ||
| XM_015113664.2 | XP_014969150.2 | 600 | 153 | |||
| XM_028826908.1 | XP_028682741.1 | 610 | 153 | |||
| GenBank mRNA | JV638227.1 | - | 485 | 153 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ITLN1
Genomic Map