Assay Details
| Gene Symbol: ZNF280C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128188.2 | XP_014983674.1 | 969 | 90 | ||
| XM_015128189.2 | XP_014983675.1 | 969 | 90 | |||
| GenBank mRNA | JU476316.1 | - | 794 | 90 | ||
More Information
Set Membership: |
|

ZNF280C
Genomic Map