Assay Details
| Gene Symbol: PIGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112393.4 | XP_001112393.2 | 2200 | 87 | ||
| XM_001112581.3 | XP_001112581.2 | 2391 | 87 | |||
| XM_015137775.2 | XP_014993261.1 | 2391 | 87 | |||
| XM_015137777.2 | XP_014993263.1 | 2627 | 87 | |||
| XM_015137778.2 | XP_014993264.1 | 2452 | 87 | |||
| XM_028848189.1 | XP_028704022.1 | 2181 | 87 | |||
| XM_028848190.1 | XP_028704023.1 | 2610 | 87 | |||
| XM_028848192.1 | XP_028704025.1 | 2581 | 87 | |||
| XM_028848193.1 | XP_028704026.1 | 1153 | 87 | |||
| GenBank mRNA | JU335137.1 | - | 2372 | 87 | ||
| JU476138.1 | - | 2383 | 87 | |||
| JU488799.1 | - | 2337 | 87 | |||
| JV047461.1 | - | 2310 | 87 | |||
More Information
Set Membership: |
|

PIGG
Genomic Map