Assay Details
| Gene Symbol: PEX26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260660.1 | NP_001247589.1 | 1042 | 65 | ||
| XM_028827328.1 | XP_028683161.1 | 885 | 65 | |||
| XM_028827329.1 | XP_028683162.1 | 946 | 65 | |||
| GenBank mRNA | JU323198.1 | - | 1042 | 65 | ||
| JU473259.1 | - | 1035 | 65 | |||
| JV639379.1 | - | 1044 | 65 | |||
More Information
Set Membership: |
|

PEX26
Genomic Map