Assay Details
| Gene Symbol: PDPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126615.2 | XP_014982101.1 | 2180 | 73 | ||
| XM_028841608.1 | XP_028697441.1 | 1952 | 73 | |||
| XM_028841609.1 | XP_028697442.1 | 2180 | 73 | |||
| XM_028841610.1 | XP_028697443.1 | 1915 | 73 | |||
| XM_028841611.1 | XP_028697444.1 | 1813 | 73 | |||
| XM_028841613.1 | XP_028697446.1 | 1615 | 73 | |||
| XM_028841614.1 | XP_028697447.1 | 2148 | 73 | |||
| XM_028841615.1 | XP_028697448.1 | 826 | 73 | |||
| GenBank mRNA | JU345194.1 | - | 1991 | 73 | ||
| JV636247.1 | - | 1958 | 73 | |||
| JV717870.1 | - | 1995 | 73 | |||
More Information
Set Membership: |
|

PDPR
Genomic Map