Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846530.1 | XP_028702363.1 | 1615 | 148 | ||
| XM_028846531.1 | XP_028702364.1 | 1573 | 148 | |||
| XM_028846532.1 | XP_028702365.1 | 1473 | 148 | |||
| XM_028846533.1 | XP_028702366.1 | 1615 | 148 | |||
| XM_028846534.1 | XP_028702367.1 | 1517 | 148 | |||
| XM_028846535.1 | XP_028702368.1 | 1019 | 148 | |||
| XM_028846536.1 | XP_028702369.1 | 442 | 148 | |||
| XM_028846537.1 | XP_028702370.1 | 961 | 148 | |||
| XM_028846538.1 | XP_028702371.1 | 863 | 148 | |||
| XM_028846539.1 | XP_028702372.1 | 755 | 148 | |||
More Information
Set Membership: |
|

DYNC2I1
Genomic Map