Assay Details
| Gene Symbol: CDCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115347.4 | XP_001115347.1 | 767 | 78 | ||
| XM_015135687.2 | XP_014991173.1 | 864 | 78 | |||
| GenBank mRNA | JU476210.1 | - | 652 | 78 | ||
More Information
Set Membership: |
|

CDCA8
Genomic Map