Assay Details
| Gene Symbol: LOC717923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111663.2 | XP_014967149.2 | 866 | 129 | ||
| GenBank mRNA | JV585475.1 | - | 129 | |||
| JV717626.1 | - | 129 | ||||
| Gene Symbol: LOC717842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116222.4 | XP_001116222.2 | 129 | |||
| XM_028828085.1 | XP_028683918.1 | 129 | ||||
| XM_028828090.1 | XP_028683923.1 | 129 | ||||
| XM_028828096.1 | XP_028683929.1 | 129 | ||||
| XM_028828100.1 | XP_028683933.1 | 129 | ||||
| XM_028828102.1 | XP_028683935.1 | 129 | ||||
| XM_028828106.1 | XP_028683939.1 | 129 | ||||
| GenBank mRNA | JV064751.1 | - | 129 | |||
More Information
Set Membership: |
|

LOC717923
Genomic Map