Assay Details
| Gene Symbol: IQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133912.2 | XP_014989398.2 | 899 | 140 | ||
| XM_015133914.2 | XP_014989400.2 | 501 | 140 | |||
| GenBank mRNA | JU345628.1 | - | 426 | 140 | ||
| JV689477.1 | - | 458 | 140 | |||
More Information
Set Membership: |
|

IQCC
Genomic Map