Assay Details
| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138470.2 | XP_014993956.1 | 1238 | 77 | ||
| XM_015138471.2 | XP_014993957.1 | 1238 | 77 | |||
| XM_015138472.2 | XP_014993958.1 | 1238 | 77 | |||
| XM_015138473.2 | XP_014993959.1 | 1302 | 77 | |||
| XM_015138474.2 | XP_014993960.1 | 1238 | 77 | |||
| XM_015138476.2 | XP_014993962.1 | 1238 | 77 | |||
| XM_015138477.2 | XP_014993963.1 | 1238 | 77 | |||
| XM_028848482.1 | XP_028704315.1 | 1254 | 77 | |||
| GenBank mRNA | JU335418.1 | - | 1221 | 77 | ||
| JU335419.1 | - | 1221 | 77 | |||
| JU472070.1 | - | 1251 | 77 | |||
| JU472071.1 | - | 1251 | 77 | |||
More Information
Set Membership: |
|

SEPTIN11
Genomic Map