Assay Details
| Gene Symbol: TMEM144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266261.1 | NP_001253190.1 | 570 | 108 | ||
| XM_015139398.2 | XP_014994884.1 | 4610 | 108 | |||
| XM_015139399.2 | XP_014994885.1 | 1132 | 108 | |||
| XM_015139400.2 | XP_014994886.1 | 1103 | 108 | |||
| XM_015139401.2 | XP_014994887.1 | 1129 | 108 | |||
| XM_015139402.2 | XP_014994888.1 | 4547 | 108 | |||
| XM_015139403.2 | XP_014994889.1 | 1100 | 108 | |||
| XM_015139404.2 | XP_014994890.1 | 1090 | 108 | |||
| XM_015139405.2 | XP_014994891.1 | 1170 | 108 | |||
| XM_028848047.1 | XP_028703880.1 | 4899 | 108 | |||
| XM_028848048.1 | XP_028703881.1 | 4544 | 108 | |||
| XM_028848049.1 | XP_028703882.1 | 4505 | 108 | |||
| XM_028848050.1 | XP_028703883.1 | 4333 | 108 | |||
| GenBank mRNA | JU322361.1 | - | 570 | 108 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM144
Genomic Map