Assay Details
| Gene Symbol: LIN7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266451.1 | NP_001253380.1 | 166 | 98 | ||
| GenBank mRNA | JU335320.1 | - | 178 | 98 | ||
| JU471953.1 | - | 165 | 98 | |||
| JU471954.1 | - | 165 | 98 | |||
| JU471955.1 | - | 165 | 98 | |||
| JU471956.1 | - | 165 | 98 | |||
| JV043503.1 | - | 166 | 98 | |||
More Information
Set Membership: |
|

LIN7C
Genomic Map