Assay Details
| Gene Symbol: LOC114675575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842790.1 | XP_028698623.1 | 202 | 91 | ||
| XM_028842791.1 | XP_028698624.1 | 202 | 91 | |||
| Gene Symbol: USE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU473791.1 | - | 165 | 91 | ||

LOC114675575
Genomic Map