Assay Details
| Gene Symbol: APOBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103862.4 | XP_001103862.2 | 962 | 93 | ||
| GenBank mRNA | JV656044.1 | - | 75 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APOBR| Gene Symbol: APOBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103862.4 | XP_001103862.2 | 962 | 93 | ||
| GenBank mRNA | JV656044.1 | - | 75 | 93 | ||
Set Membership: |
|