Assay Details
| Gene Symbol: ADARB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001118272.4 | XP_001118272.3 | 1753 | 138 | ||
| GenBank mRNA | JV603572.1 | - | 777 | 138 | ||
| JV670633.1 | - | 548 | 138 | |||
More Information
Set Membership: |
|

ADARB2
Genomic Map