Assay Details
| Gene Symbol: CTTNBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257547.1 | NP_001244476.1 | 511 | 85 | ||
| XM_015147066.2 | XP_015002552.1 | 716 | 85 | |||
| GenBank mRNA | JU321676.1 | - | 511 | 85 | ||
| JV044086.1 | - | 525 | 85 | |||
More Information
Set Membership: |
|

CTTNBP2NL
Genomic Map