Assay Details
| Gene Symbol: C10H20orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190968.1 | NP_001177897.1 | 215 | 89 | ||
| XM_028827324.1 | XP_028683157.1 | 501 | 89 | |||
| GenBank mRNA | CK230208.1 | - | 77 | 89 | ||
| JU320668.1 | - | 355 | 89 | |||
| JU476412.1 | - | 273 | 89 | |||
| JV047652.1 | - | 401 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C10H20orf24
Genomic Map