Assay Details
| Gene Symbol: TUBA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104173.4 | XP_001104173.1 | 927 | 72 | ||
| XM_015149685.2 | XP_015005171.1 | 519 | 72 | |||
| GenBank mRNA | JU328141.1 | - | 158 | 72 | ||
| JV716832.1 | - | 328 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015149685.2 transcripts.
Set Membership: |
|

TUBA8
Genomic Map