Assay Details
| Gene Symbol: GNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265767.1 | NP_001252696.1 | 725 | 103 | ||
| XM_015136362.2 | XP_014991848.1 | 958 | 103 | |||
| GenBank mRNA | JU484540.1 | - | 618 | 103 | ||
| JV635765.1 | - | 725 | 103 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GNMT
Genomic Map