Assay Details
| Gene Symbol: TREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001117305.4 | XP_001117305.2 | 613 | 96 | ||
| XM_015136314.2 | XP_014991800.1 | 942 | 96 | |||
| GenBank mRNA | JU335059.1 | - | 580 | 96 | ||
| JV510268.1 | - | 382 | 96 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TREM2
Genomic Map