Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147056.2 | XP_015002542.2 | 1647 | 150 | ||
XM_028826364.1 | XP_028682197.1 | 1588 | 150 | |||
XM_028826365.1 | XP_028682198.1 | 1588 | 150 | |||
XM_028826366.1 | XP_028682199.1 | 1588 | 150 | |||
XM_028826367.1 | XP_028682200.1 | 1588 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() |
