Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147056.2 | XP_015002542.2 | 1757 | 127 | ||
XM_028826364.1 | XP_028682197.1 | 1698 | 127 | |||
XM_028826365.1 | XP_028682198.1 | 1698 | 127 | |||
XM_028826366.1 | XP_028682199.1 | 1698 | 127 | |||
XM_028826368.1 | XP_028682201.1 | 561 | 127 | |||
XM_028826369.1 | XP_028682202.1 | 188 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028826369.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
