Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150480.2 | XP_015005966.1 | 778 | 82 | ||
| XM_015150481.2 | XP_015005967.1 | 803 | 82 | |||
| XM_015150482.2 | XP_015005968.1 | 866 | 82 | |||
| XM_028827647.1 | XP_028683480.1 | 2066 | 82 | |||
| XM_028827648.1 | XP_028683481.1 | 2066 | 82 | |||
| XM_028827649.1 | XP_028683482.1 | 823 | 82 | |||
| XM_028827650.1 | XP_028683483.1 | 984 | 82 | |||
| GenBank mRNA | JU319811.1 | - | 686 | 82 | ||
| JV587371.1 | - | 697 | 82 | |||
| JV587372.1 | - | 697 | 82 | |||
More Information
Set Membership: |
|

SEPTIN3
Genomic Map