Assay Details
| Gene Symbol: ALG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125564.2 | XP_014981050.1 | 1108 | 67 | ||
| XM_015125565.2 | XP_014981051.1 | 1108 | 67 | |||
| XM_015125567.2 | XP_014981053.1 | 916 | 67 | |||
| XM_028841009.1 | XP_028696842.1 | 1253 | 67 | |||
| GenBank mRNA | JU325402.1 | - | 1039 | 67 | ||
| JU472924.1 | - | 1096 | 67 | |||
More Information
Set Membership: |
|

ALG1
Genomic Map