Assay Details
| Gene Symbol: NDUFA4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115919.4 | XP_001115919.1 | 1137 | 95 | ||
| XM_028829656.1 | XP_028685489.1 | 1144 | 95 | |||
| GenBank mRNA | JU325648.1 | - | 208 | 95 | ||
| JV656376.1 | - | 207 | 95 | |||
More Information
Set Membership: |
|

NDUFA4L2
Genomic Map