Assay Details
| Gene Symbol: DTWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260730.1 | NP_001247659.1 | 720 | 153 | ||
| XM_015142229.2 | XP_014997715.2 | 864 | 153 | |||
| XM_015142232.2 | XP_014997718.2 | 867 | 153 | |||
| XM_028850495.1 | XP_028706328.1 | 1242 | 153 | |||
| XM_028850496.1 | XP_028706329.1 | 723 | 153 | |||
| GenBank mRNA | JU322502.1 | - | 807 | 153 | ||
| JU476475.1 | - | 720 | 153 | |||
| JV047713.1 | - | 785 | 153 | |||
| JV656460.1 | - | 810 | 153 | |||
More Information
Set Membership: |
|

DTWD1
Genomic Map