Assay Details
| Gene Symbol: PAK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260987.1 | NP_001247916.1 | 1791 | 146 | ||
| XM_015149385.2 | XP_015004871.1 | 1963 | 146 | |||
| XM_028827454.1 | XP_028683287.1 | 1784 | 146 | |||
| GenBank mRNA | JU321969.1 | - | 1791 | 146 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PAK5
Genomic Map