Assay Details
| Gene Symbol: NKIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266422.2 | NP_001253351.1 | 116 | 140 | ||
| XM_015132252.2 | XP_014987738.1 | 246 | 140 | |||
| XM_015132253.2 | XP_014987739.1 | 251 | 140 | |||
| XM_015132254.2 | XP_014987740.1 | 273 | 140 | |||
| XM_015132256.2 | XP_014987742.1 | 358 | 140 | |||
| XM_015132258.2 | XP_014987744.1 | 253 | 140 | |||
| XM_028843158.1 | XP_028698991.1 | 134 | 140 | |||
| GenBank mRNA | JU322245.1 | - | 120 | 140 | ||
| JV465655.1 | - | 143 | 140 | |||
| JV503674.1 | - | 119 | 140 | |||
More Information
Set Membership: |
|

NKIRAS1
Genomic Map