Assay Details
| Gene Symbol: FSCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089987.4 | XP_001089987.2 | 1699 | 94 | ||
| XM_015134847.2 | XP_014990333.2 | 1651 | 94 | |||
| XM_015134849.2 | XP_014990335.2 | 1989 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FSCN3
Genomic Map