Assay Details
| Gene Symbol: EIF5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260939.1 | NP_001247868.1 | 84 | 120 | ||
| GenBank mRNA | JU335502.1 | - | 84 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260939.1 transcripts.
Set Membership: |
|

EIF5A2
Genomic Map