Assay Details
| Gene Symbol: PELI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260952.1 | NP_001247881.1 | 798 | 121 | ||
| XM_028831440.1 | XP_028687273.1 | 1230 | 121 | |||
| XM_028831441.1 | XP_028687274.1 | 858 | 121 | |||
| GenBank mRNA | JU319988.1 | - | 1064 | 121 | ||
| JU476500.1 | - | 1034 | 121 | |||
| JV045492.1 | - | 798 | 121 | |||
More Information
Set Membership: |
|

PELI1
Genomic Map