Assay Details
| Gene Symbol: MRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100770.4 | XP_001100770.2 | 316 | 175 | ||
| XM_002803622.3 | XP_002803668.2 | 332 | 175 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRS2| Gene Symbol: MRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100770.4 | XP_001100770.2 | 316 | 175 | ||
| XM_002803622.3 | XP_002803668.2 | 332 | 175 | |||
Set Membership: |
|