Assay Details
| Gene Symbol: NUFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261811.1 | NP_001248740.1 | 289 | 75 | ||
| GenBank mRNA | JU335643.1 | - | 346 | 75 | ||
| JU476544.1 | - | 289 | 75 | |||
| JU476545.1 | - | 289 | 75 | |||
More Information
Set Membership: |
|

NUFIP2
Genomic Map