Assay Details
| Gene Symbol: GBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105060.4 | XP_001105060.2 | 160 | 67 | ||
| XM_028848368.1 | XP_028704201.1 | 160 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GBA3| Gene Symbol: GBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105060.4 | XP_001105060.2 | 160 | 67 | ||
| XM_028848368.1 | XP_028704201.1 | 160 | 67 | |||
Set Membership: |
|