Assay Details
| Gene Symbol: AQP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094912.4 | XP_001094912.1 | 740 | 83 | ||
| XM_028850982.1 | XP_028706815.1 | 740 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AQP9
Genomic Map