Assay Details
| Gene Symbol: DEAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113248.2 | XP_014968734.1 | 2347 | 71 | ||
| XM_028832984.1 | XP_028688817.1 | 1667 | 71 | |||
| GenBank mRNA | JV049776.1 | - | 3366 | 71 | ||
| JV603632.1 | - | 1522 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DEAF1
Genomic Map